Learn More
Abeba™ Reflexor™ 36902 Shoes
Product Code. 15600798
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
Unit Size:
1 pair
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15600798 36 1 pair
15610798 37 1 pair
15620798 38 1 pair
15630798 39 1 pair
15640798 40 1 pair
15650798 41 1 pair
15660798 42 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 15600798 Supplier Abeba™ Supplier No. 36902/36
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
ESD dissipative shoe includes PU-sole with high slip resistance.
  • CE EN ISO 20347:2012 OB A, E SRB
  • Smooth leather white
  • Slip resistance PU outsole sole
  • 2 replaceable Reflexor™ insoles (article no. 3565/ 3567)
  • ESD dissipative according to DIN EN 61340
  • Lining microfiber, respiration absorbing
  • 3 adjustable instep straps
  • velcro fastening
  • Hand-sewn seam
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E SRB
Color White
Liner Material Microfiber
Material Leather
Size 36
Sole Features Slip-resistant
Sole Material Polyurethane
Style 36902