Learn More
Labcon Refillable Racks for Labcon Tips
For use with Eclipse™ pipette tip refills
Brand:  Labcon 1299-960-036-9
Additional Details : Weight : 11.33980kg
 View more versions of this product
Product Code. 16681963
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Pipette tip rack
  • Refillable
  • With hinged clear lid
  • Made in USA
Specifications
Specifications
Refillable
Autoclavable
Tips (Labcon 10μL)
Holds one refill platform of 96 tips
Liquid Handling
12 racks / Pk.
10 μL tips
ISO 9001:2015; FDA CFR21 GMP
Purple
Polypropylene
Non-sterile
12 racks / Pk./10 Pk. / case
RT