missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Non-magnetizable
- E2 weights made of special stainless steel with a density of 8,000 kg/m³
- F1 weights made of special stainless steel with a density of 7,950 kg/m³
- With DAkkS calibration certificate
Specifications
Specifications
| Product Type | Reference Line Weight |
| Certifications/Compliance | DAkkS calibration certificate |
| Classification | F1 |
| Material | Stainless Steel |
| Weight (Metric) | 500 mg |
| Density | 7,950 kg/m³ |
| For Use With (Application) | Primary mass standards |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?