missing translation for 'onlineSavingsMsg'
Learn More

Reference Gas Oil No. 2 for ASTM D2887

Product Code. 30322546
Click to view available options
Quantity:
10 x 1 mL
2 fl.oz.
Unit Size:
10mL
Pack of 2
This item is not returnable. View return policy

Product Code. 30322546

Brand: VHG Labs VHGRGO2OZ

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Shipping Condition Room Temperature
Solution Type Single Solution
Solvent or Matrix Reference Gas Oil
Industry Type Petrochemical Testing
Quantity 2 fl.oz.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.