missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ REEP6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106432
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total REEP6.
REEP6 may enhance the cell surface expression of odorant receptors.
Specifications
| REEP6 | |
| Polyclonal | |
| Unconjugated | |
| REEP6 | |
| 0610011M24Rik; C19orf32; deleted in polyposis 1-like 1; DP1L1; Polyposis locus protein 1-like 1; receptor accessory protein 6; receptor expression enhancing protein 6; receptor expression-enhancing protein 6; REEP6; TB2 protein-like 1; Tb2l1; Yip2f | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 362835, 70335, 92840 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5XI60, Q96HR9, Q9JM62 | |
| REEP6 | |
| A synthesized peptide derived from human REEP6(Accession Q96HR9), corresponding to amino acid residues R181-K211. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction