missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ REEP5 Monoclonal Antibody (OTI4D2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526063
This item is not returnable.
View return policy
Description
REEP5 Monoclonal Antibody for Western Blot, IHC (P), Flow
REEP5 (receptor expression-enhancing protein 5), also known as C5orf18, DP1, TB2 or D5S346, is a 189 amino acid multi-pass membrane protein. Thought to promote the functional cell surface expression of olfactory receptors, REEP5 belongs to the DP1 family and is encoded by a gene that maps to chromosome 5. With 181 million base pairs encoding around 1,000 genes, chromosome 5 is about 6% of human genomic DNA. Chromosome 5 is associated with Cockayne syndrome through the ERCC8 gene and familial adenomatous polyposis through the adenomatous polyposis coli (APC) tumor suppressor gene. Treacher Collins syndrome is also chromosome 5 associated and is caused by insertions or deletions within the TCOF1 gene. Deletion of the parm of chromosome 5 leads to cri du chat syndrome. Deletion of 5q or chromosome 5 altogether is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.
Specifications
| REEP5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q00765 | |
| REEP5 | |
| Full length human recombinant protein of REEP5 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI4D2 | |
| Unconjugated | |
| REEP5 | |
| AI324241; AU022809; AW495741; C5orf18; D5S346; deleted in polyposis 1; Dp1; DP1/TB2; GP106; polyposis coli region hypothetical protein DP1; polyposis locus protein 1; Polyposis locus protein 1 homolog; Protein TB2; protein TB2 homolog; R74856; receptor accessory protein 5; receptor expression enhancing protein 5; receptor expression-enhancing protein 5; Reep5; TB2; TB2/DP1; Yip2e; YOP1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7905 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction