missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ REEP4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12849032
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12849032 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12849032 Supplier Invitrogen™ Supplier No. PA520885

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is human lung tissue lysate. PA5-20885 can be used with blocking peptide PEP-0999.

Microtubule-binding protein required to ensure proper cell division and nuclear envelope reassembly by sequestering the endoplasmic reticulum away from chromosomes during mitosis. Probably acts by clearing the endoplasmic reticulum membrane from metaphase chromosomes. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen REEP4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene REEP4
Gene Accession No. Q4QQW1, Q8K072, Q9H6H4
Gene Alias 2700029E10Rik; C8orf20; PP432; receptor accessory protein 4; receptor expression enhancing protein 4; receptor expression-enhancing protein 4; Reep4; RGD1306561; Yip2c
Gene Symbols REEP4
Host Species Rabbit
Immunogen A 17 amino acid peptide near the center of human REEP4.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 306014, 72549, 80346
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.