missing translation for 'onlineSavingsMsg'
Learn More

REEP4 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18409086 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409086 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409086 Supplier Novus Biologicals Supplier No. NBP176255JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

REEP4 Polyclonal antibody specifically detects REEP4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen REEP4
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias C8orf20chromosome 8 open reading frame 20, FLJ22246, FLJ22277, PP432, receptor accessory protein 4, receptor expression enhancing protein 4, receptor expression-enhancing protein 4
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the center of human REEP4. The immunogen is located within amino acids 160 - 210 of REEP4. Amino Acid Squence: AQRWQPGSRGEKVTGGY
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 80346
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.