missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REEP4 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-30361F
This item is not returnable.
View return policy
Description
REEP4 Polyclonal antibody specifically detects REEP4 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)
Specifications
| REEP4 | |
| Polyclonal | |
| Immunoprecipitation, Western Blot (Negative) | |
| C8orf20chromosome 8 open reading frame 20, FLJ22246, FLJ22277, PP432, receptor accessory protein 4, receptor expression enhancing protein 4, receptor expression-enhancing protein 4 | |
| Maps to a region between residue 200 to 225 of human Receptor Accessory Protein 4 using the numbering given in entry NP_079508.2 (GeneID 80346). | |
| 0.1 mL | |
| Signal Transduction | |
| 80346 | |
| Store at 4C in the dark. | |
| IgG |
| Immunoprecipitation | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction