Learn More
Invitrogen™ REEP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA520884
Product Code. 12839032
  • £440.00 / 100µg
Description
A suggested positive control is rat lung tissue lysate. PA5-20884 can be used with blocking peptide PEP-0998.
Mammalian odorant receptors require accessory proteins such as RTP1 and RTP2 for functional cell surface expression. Receptor expression-enhancing protein (REEP) family members are transmembrane proteins which interact with odorant receptor proteins and may enhance the odorant receptor responses to odorants. Recently studies have shown other chemosensory receptors such as bitter taste receptors are also influenced by RTP and REEP family members. REEP3 has also been suggested as a novel candidate gene for autism and as a loci influencing the plasma levels of the liver enzyme alkaline phosphatase.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
REEP3
Polyclonal
Unconjugated
REEP3
C10orf74; D10Ucla1; receptor accessory protein 3; receptor expression enhancing protein 3; receptor expression-enhancing protein 3; REEP3; RGD1560401; RP11-439F7.1; Yip2b
Rabbit
Antigen affinity chromatography
RUO
221035, 28193, 294375
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Immunohistochemistry, Western Blot, Immunocytochemistry
1 mg/mL
PBS with 0.02% sodium azide
Q6NUK4, Q99KK1
REEP3
A 17 amino acid peptide near the carboxy terminus of human REEP3.
100 μg
Primary
Human, Mouse, Rat