missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REEP3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£134.00
Specifications
| Antigen | REEP3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
Description
REEP3 Polyclonal specifically detects REEP3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| REEP3 | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| NP_001001330 | |
| 221035 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human REEP3. The immunogen is located within the last 50 amino acids of REEP3. Amino Acid Squence: GRKEVRYGSLKYKVKKR | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| C10orf74receptor expression enhancing protein 3, chromosome 10 open reading frame 74, MGC87479, receptor accessory protein 3, receptor expression-enhancing protein 3 | |
| REEP3 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title