Learn More
Invitrogen™ REEP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113912
Description
Immunogen sequence: ERLRSFSMQD LTTIRGDGAP APSGPPPPGS GRASGKHGQP KMSRSASESA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 92%; Dog: 86%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
REEP1 belongs to the DP1 family and may enhance the cell surface expression of odorant receptors.
Specifications
| REEP1 | |
| Polyclonal | |
| Unconjugated | |
| Reep1 | |
| C2orf23; D6Ertd253e; HMN5B; rcG_56072; receptor accessory protein 1; receptor expression-enhancing protein 1; receptor expression-enhancing protein 1-like; Reep1; RGD1305230; si:dkey-21a6.4; Spastic paraplegia 31 protein; SPG31; Yip2a | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 65055 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H902 | |
| Reep1 | |
| Synthetic peptide directed towards the C terminal region of human REEP1 (aa 146-195). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.