missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ REEP1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15576042
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15576042 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15576042 Supplier Invitrogen™ Supplier No. PA543424

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: AKDRSYDALV HFGKRGLNVA ATAAVMAASK GQGALSERLR SFSMQDLTTI Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.

REEP1 belongs to the DP1 family and may enhance the cell surface expression of odorant receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REEP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Reep1
Gene Accession No. Q9H902
Gene Alias C2orf23; D6Ertd253e; HMN5B; rcG_56072; receptor accessory protein 1; receptor expression-enhancing protein 1; receptor expression-enhancing protein 1-like; Reep1; RGD1305230; si:dkey-21a6.4; Spastic paraplegia 31 protein; SPG31; Yip2a
Gene Symbols Reep1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human REEP1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65055
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.