missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reelin Antibody (RELN/4498) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14203JF585
This item is not returnable.
View return policy
Description
Reelin Monoclonal antibody specifically detects Reelin in Human,Mammalia,Mouse samples. It is validated for ELISA,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Reelin | |
| Monoclonal | |
| Janelia Fluor 585 | |
| EC 3.4.21, EC 3.4.21.-, reelin, RLPRO1598 | |
| Recombinant fragment (around aa 1796-2115) of human Reelin protein (Uniprot: P78509) | |
| 0.1 mL | |
| Primary | |
| Human, Mammalia, Mouse | |
| Purified |
| ELISA, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| RELN/4498 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 5649 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction