Learn More
Lenz Laborglasintrumente Reduction Adapters with Ground Joints
Product Code. 10060201
Change view
Click to view available options
Standard Taper Joint No.:
Socket NS 14/23
Socket NS 19/26
Socket NS 24/29
Socket NS 29/32
Socket NS 34/35
Socket NS 45/40
Cone Size:
NS 19/26
NS 24/29
NS 29/32
NS 34/35
NS 45/40
NS 60/46
NS 71/51
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No. Cone Size unitSize
10060201 Socket NS 34/35 NS 19/26 Each
10002933 Socket NS 45/40 NS 71/51 Each
10707331 Socket NS 34/35 NS 29/32 Each
10070351 Socket NS 29/32 NS 71/51 Each
10224051 Socket NS 45/40 NS 60/46 Each
10274571 Socket NS 24/29 NS 45/40 Each
10284331 Socket NS 19/26 NS 34/35 Each
10385451 Socket NS 45/40 NS 34/35 Each
10684192 Socket NS 34/35 NS 24/29 Each
10517701 Socket NS 14/23 NS 29/32 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 10060201 Supplier Lenz Laborglasintrumente Supplier No. 01503419
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Acc. to DIN 12257 and in addition to DIN standard
  • Conical ground sockets and cones
  • Made of DURAN™ tubing
Specifications
Cone Size NS 19/26
Material Borosilicate Glass (3.3)
Standard Taper Joint No. Socket NS 34/35