missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reduced Folate Carrier/SLC19A1 Antibody (890513), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8450CL1
This item is not returnable.
View return policy
Description
Reduced Folate Carrier/SLC19A1 Monoclonal antibody specifically detects Reduced Folate Carrier/SLC19A1 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| Reduced Folate Carrier/SLC19A1 | |
| Monoclonal | |
| CoraFluor 1 | |
| CHMD, FLOT1, folate transporter 1, FOLThuman reduced folate carrier (RFC)10Intestinal folate carrier 1, IFC1, IFC-1, Placental folate transporter, Reduced folate carrier protein, REFC, RFC, RFC1, solute carrier family 19 (folate transporter), member 1, Solute carrier family 19 member 1 | |
| NS0 mouse myeloma cell line transfected with human Reduced Folate Carrier/SLC19A1, Met1-Gln591, Accession # P41440 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 890513 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 6573 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction