missing translation for 'onlineSavingsMsg'
Learn More

SI Analytics™ Redox Solution, 430mV Pt/Calomel, 470mV Pt/Ag/AgCl

Product Code. p-8014346 Shop All SI Analytics Products
Click to view available options
Quantity:
1000 mL
60 Ampules/20mL
Unit Size:
1000mL
Each
This item is not returnable. View return policy

Product Code. 10664471

Brand: SI Analytics™ 285138184

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SI Analytics™ Redox Solution, 430mV Pt/Calomel, 470mV Pt/Ag/AgCl is designed to be used in redox tests.

  • GHS07: Skin Irrit. 2 H315 Causes skin irritation, Eye Irrit. 2 H319 Causes serious eye irritation
  • Precautionary statements: P280 Wear protective gloves / eye protection / face protection. P264 Wash thoroughly after handling. P305+P351+P338 IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. P321 Specific treatment (see on this label). P332+P313 If skin irritation occurs: Get medical advice/attention. P337+P313 If eye irritation persists: Get medical advice/attention. P302+P352 IF ON SKIN: Wash with plenty of water.
  • Ingredients with limit values that require monitoring at the workplace: 7664-93-9 sulphuric acid
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 102°C
Color Light Yellow
Conductivity 430 Pt/Kalomel, 470 Pt/Ag/Agcl
Filling Solution Type Redox
Odor Odorless
Packaging DURAN™ Bottle
pH Range -0.1
Quantity 1000 mL
Solubility Fully miscible in water
Vapor Pressure 23 hPa
compliance-icons compliance-icons
Product Identifier
  • Redox testing solution
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.