missing translation for 'onlineSavingsMsg'
Learn More

SI Analytics™ Redox Solution, 430mV Pt/Calomel, 470mV Pt/Ag/AgCl

Product Code. 10340641 Shop All SI Analytics Products
Change view
Click to view available options
Quantity:
1000 mL
60 Ampules/20mL
Unit Size:
1000mL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10340641 60 Ampules/20mL Each
10664471 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10340641 Supplier SI Analytics™ Supplier No. 285138357

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SI Analytics™ Redox Solution, 430mV Pt/Calomel, 470mV Pt/Ag/AgCl is designed to be used in redox tests.

  • GHS07: Skin Irrit. 2 H315 Causes skin irritation, Eye Irrit. 2 H319 Causes serious eye irritation
  • Precautionary statements: P280 Wear protective gloves / eye protection / face protection. P264 Wash thoroughly after handling. P305+P351+P338 IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. P321 Specific treatment (see on this label). P332+P313 If skin irritation occurs: Get medical advice/attention. P337+P313 If eye irritation persists: Get medical advice/attention. P302+P352 IF ON SKIN: Wash with plenty of water.
  • Ingredients with limit values that require monitoring at the workplace: 7664-93-9 sulphuric acid
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 102°C
Color Light Yellow
Conductivity 430 Pt/Kalomel, 470 Pt/Ag/AgclMv
Filling Solution Type Redox
Odor Odorless
Packaging FIOLAX™ Ampules
pH Range -0.1
Quantity 60 Ampules/20mL
Solubility Fully miscible in water
Vapor Pressure 23 hPa
compliance-icons
Product Identifier
  • Redox test solution (+430 mV+470 mV)
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P362-Take off contaminated clothing.
  • P364-And wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: sulphuric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.