missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REDD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10638-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is a rabbit polyclonal antibody raised against full length human REDD1 antigen. This antibody is specific to the REDD1 from siRNA experiment.
DDIT4 gene ontology annotations related to this gene include cellular response to dexamethasone stimulus; intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator; negative regulation of glycolytic process; negative regulation of TOR signaling; protein-containing complex disassembly; reactive oxygen species metabolic process; response to hypoxia.Specifications
| REDD1 | |
| Polyclonal | |
| Unconjugated | |
| DDIT4 | |
| DDIT4, Dig2, REDD 1, REDD1, RP11 442H21.1, RTP801 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 140942, 54541, 74747 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.53 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8VHZ9, Q9D3F7, Q9NX09 | |
| DDIT4 | |
| REDD1 specific Fusion Protein Ag0965 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction