missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Redcote Plus Gloves

Catalog Number 15668655
Click to view available options
Size:
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Suitable for handling objects in an aggressive environment of oils, greases, chemicals and hydrocarbons. Honeywell Safety Products™ Redcote Plus Gloves are a sturdy multipurpose glove with a cotton interlock lining for comfort and a smooth finish for touch and dexterity.

  • Handling in an aggressive environment of oils, greases, chemicals, hydrocarbons.
  • Suitable for chemical and petrochemical industries.
  • Cotton interlock lining absorbs hand perspiration for added comfort and adds to mechanical performance.
  • Articulated fingers offer superior comfort and reduce effort in cases extended use.
  • PVC composition tailored for multi-purpose use and repetitive handling in an oily atmosphere.
  • Low resistance to ketones, aromatic and halogen hydrocarbons.
  • Cuff helps to protect a large part of the forearm.
TRUSTED_SUSTAINABILITY
Certifications/Compliance EC Category PPE 3, EC Certificate Number 0075/797/162/05/13/0457 EXT 02/05/13, EC Attestation Number 0075/797/162/05/13/0457 EXT 02/05/13, EN 0075/797/162/05/13/0457 EXT 02/05/13
Classification Dexterity: 5
Coated Area Exterior
Coating Material PVC
Color Red
Cut Resistance Rating 1
Exterior Finish Smooth
Material PVC, Cotton
Product Type Gloves
Size 10
For Use With (Application) Agriculture, Chemical, Petrochemical and Water Treatment industries.
Liner Material Cotton
Lining Lined
Puncture Rating 1
Quantity 12Pair/PK
Tear Rating 2
Length (Metric) 40 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'