missing translation for 'onlineSavingsMsg'
Learn More

RED1 Sheep anti-Rat, Polyclonal, Novus Biologicals™

Product Code. 18342842 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 μg
50 μg
Unit Size:
25µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342842 25 μg 25µg
18309822 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18342842 Supplier Novus Biologicals Supplier No. NBP31188725UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

RED1 Polyclonal antibody specifically detects RED1 in Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RED1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS (0.2 micron sterile filtered solution)
Gene Alias ADAR2a, ADAR2ADAR2g, ADAR2a-L1, ADAR2a-L2, ADAR2a-L3, ADAR2b, ADAR2c, adenosine deaminase, RNA-specific, B1, double-stranded RNA-specific editase 1, DRABA2, DRADA2hRED1, EC 3.5, human dsRNA adenosine deaminase DRADA2, human dsRNA adenosine deaminase DRADA2b, EC 3.510adenosine deaminase, RNA-specific, B1 (homolog of rat RED1), RED1 homolog, RED1ADAR2d, RNA editase, RNA editing deaminase 1, RNA-editing deaminase 1, RNA-editing enzyme 1, RNA-specific, B1 (RED1 homolog rat)
Host Species Sheep
Immunogen Sheep were immunized with a GST fusion protein containing amino acid sequence 6-66 of rat RED1 (Uniprot: P51400)
Purification Method Ammonium sulfate precipitation
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104
Target Species Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.