missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RED Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101789
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RED.
Insulin-like growth factor-II messenger RNA (mRNA)-binding protein-3 (IMP-3), also known as K homology domain-containing protein overexpressed in cancer (KOC) and L523S, is a member of the insulin-like growth factor-II mRNA-binding protein family and is expressed during embryogenesis and in some malignancies. IMP-3 is expressed in malignant melanoma but not in benign nevi, even when dysplastic features are present; IMP-3 is expressed in a significantly higher proportion of melanomas than Spitz nevi; and IMP-3 is expressed in metastatic melanomas significantly more than in thin melanomas. IMP-3 appears to be involved in the progression of malignant melanoma and may play an important role in the regulation of the biologic behavior of this tumor.
Specifications
| RED | |
| Polyclonal | |
| Unconjugated | |
| IK | |
| arginine/glutamic acid/aspartic acid repeat containing protein; chondrosarcoma-associated protein 2; CSA2; Cytokine IK; Cytokine IK factor; IK; IK cytokine; IK cytokine, down-regulator of HLA II; IK factor; MuRED; prer protein; protein Red; Protein RER; RD element; RED; Rer | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 24010, 291659, 3550 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13123, Q66HG8, Q9Z1M8 | |
| IK | |
| A synthesized peptide derived from human IK(Accession Q13123), corresponding to amino acid residues R518-Y557. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction