missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Red Nucleic Acid Gel Stain (10,000x), MedChemExpress
Description
Red Nucleic Acid Gel Stain (10,000x) is a nucleic acid stain that can be used as a safer alternative to the traditional ethidium bromide (EB) stain for detecting nucleic acids in agarose gels or polyacrylamide gels.
Specifications
Specifications
| Chemical Name or Material | Red Nucleic Acid Gel Stain (10,000x) |
| Physical Form | Liquid |
| Color | Reddish Brown |
| Quantity | 500 μL |
| Purity Grade Notes | Research |
| Grade | Research |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?