Learn More
Melford Red-GlcA
Product Code. 30018204
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
30018204 100 mg 100mg
1 options
Product Code. 30018204 Supplier Melford Supplier No. C730600.1
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Chromogenic substrate for β-D-Glucuronidase (GUS).
Chromogenic substrate for β-D-Glucuronidase (GUS). Produces a red color for (GUS) gene detection.
Specifications
Quantity 100 mg