Learn More
Thermo Scientific™ pCMV-Red Firefly Luc Vector for Luciferase Assays
Codice prodotto. 12102240 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Quantity:
10 μg
Dimensione della confezione:
10µg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 12102240
Marca: Thermo Scientific™ 16156
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Drive red-shifted firefly luciferase expression as a reporter for gene regulation studies using vectors with a multiple cloning site (MCS) or control promoter (CMV or TK).
The Thermo Scientific™ pMCS-Red Firefly Luc Vector is a multiple cloning site plasmid designed to accept a promoter sequence for study of gene regulation using the intracellular red firefly luciferase reporter.
The Red Firefly Luc Vectors contain a mutant form of the firefly luciferase gene that has a red-shifted emission spectrum. The pMCS vector contains a multiple cloning site for cloning a promoter to study its regulatory potential. The pCMV and pTK vectors have the luciferase gene under the CMV (Cytomegalovirus) promoter and Herpes Simplex Virus (HSV) thymidine kinase (TK) promoter, respectively. These constitutive expression vectors can be used as normalization controls to account for experimental variation in combination with other reporters.
Highlights:
  • Intracellular Red Firefly Luciferase gene, optimized for high expression in mammalian systems
  • Multiple cloning site (MCS) provides versatility for transfer of regulatory elements from one plasmid to another
  • Transcription termination site (Ter), Lac operator (Lac O1), and transcriptional pause site (TPS) used to minimize background by reducing transcriptional read-through
  • Both puromycin (Pur) and ampicillin (Amp) markers for drug selection in mammalian and bacterial cells, respectively
  • High-copy pUC bacterial DNA replication origin
  • Two control vectors available with strong (CMV) or weak (TK) constitutive promoters for co-transfection and normalization
TRUSTED_SUSTAINABILITY
Specifica
Description pCMV-Red Firefly Luc Vector
Type Luc Vector
Cloning Method Restriction Enzyme/MCS
Delivery Type Transfection
Quantity 10 μg
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: -70 to -20°C.
Reporter Gene Red Firefly Luciferase
Selection Agent (Eukaryotic) Puromycin
Product Line Pierce
For Use With (Application) Reporter Assays
Promoter CMV
Vedi altri risultati Mostra meno risultati
For Research Use Only. Not for use in diagnostic procedures.
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado