missing translation for 'onlineSavingsMsg'
Learn More

Red Blood Cells Antibody (34-3C), Novus Biologicals™

Product Code. 18392003 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18392003 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18392003 Supplier Novus Biologicals Supplier No. NBP311200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Red Blood Cells Monoclonal specifically detects Red Blood Cells in Mouse samples. It is validated for Flow Cytometry, Functional.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Red Blood Cells
Applications Flow Cytometry, Functional Assay
Classification Monoclonal
Clone 34-3C
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry, Functional
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias Erythrocytes, Erythroid Cells, Haematids, RBC
Host Species Mouse
Immunogen RBC
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.