Learn More
Gilac™ Rectangular Container
Product Code. 17918541
Change view
Click to view available options
Volume (Metric):
12 L
15 L
25 L
35 L
55 L
Color:
Gray
White
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Color unitSize
17918541 35 L White Each
17918621 12 L White Each
17958851 15 L White Each
17928531 15 L Gray Each
17948931 25 L Gray Each
17958541 35 L Gray Each
17988481 55 L Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 17918541 Supplier Gilac™ Supplier No. G119821
Only null left
Add to basket
Add to basket
Rectangular Container is for sub-zero and ambient temperature storage and transporting raw materials.
  • Rectangular plastic food container.
  • For sub-zero and ambient temperature storage and transporting raw materials.
  • No gaps nor overlaps.
  • Machine washable.
  • Covering lid for increased hygiene, optional extra.
  • Stackable without lid.
  • Compatible with all GILAC 4 and 6-wheel dollies.
Specifications
Product Type Food Container
Material Polyethylene
Color White
Shape Rectangular
Volume (English) 9.2 gal.
Volume (Metric) 35 L