missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RECQL5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12468-2-AP-20UL
This item is not returnable.
View return policy
Description
RECQL5 gene ontology annotations related to this gene include cellular response to camptothecin; chromosome separation; DNA duplex unwinding; DNA recombination; DNA repair; DNA replication; double-strand break repair via homologous recombination; mitotic sister chromatid segregation; negative regulation of double-strand break repair via homologous recombination; negative regulation of mitotic cell cycle; negative regulation of transcription elongation from RNA polymerase II promoter; replication-born double-strand break repair via sister chromatid exchange; response to X-ray.Specifications
| RECQL5 | |
| Polyclonal | |
| Unconjugated | |
| RECQL5 | |
| ATP dependent DNA helicase Q5, DNA helicase, RecQ like type 5, RecQ protein like 5, RECQ5, RECQL5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 170472, 287834, 9400 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| D4ACP5, O94762, Q8VID5 | |
| Recql5 | |
| RECQL5 Fusion Protein Ag3145 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido