Learn More
Invitrogen™ RECQL5 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5115832
Product Code. 17248613
  • £452.00 / 100µL
Description
Antibody detects endogenous levels of total RECQL5.
RECQL5 gene ontology annotations related to this gene include cellular response to camptothecin; chromosome separation; DNA duplex unwinding; DNA recombination; DNA repair; DNA replication; double-strand break repair via homologous recombination; mitotic sister chromatid segregation; negative regulation of double-strand break repair via homologous recombination; negative regulation of mitotic cell cycle; negative regulation of transcription elongation from RNA polymerase II promoter; replication-born double-strand break repair via sister chromatid exchange; response to X-ray.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RECQL5
Polyclonal
Unconjugated
RECQL5
ATP-dependent DNA helicase Q5; DNA helicase, RecQ-like type 5; FLJ90603; LOW QUALITY PROTEIN: ATP-dependent DNA helicase Q5; RecQ helicase-like 5; RecQ like helicase 5; RecQ protein 5; RecQ protein-like 5; RecQ protein-like 5 like; RECQ5; Recq5b; RECQL5; Recql5b; RECQL5beta; recql5l
Rabbit
Affinity Chromatography
RUO
170472, 287834, 9400
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide
D4ACP5, O94762, Q8VID5
RECQL5
A synthesized peptide derived from human RECQL5(Accession O94762), corresponding to amino acid residues E808-S858.
100 μL
Primary
Human, Mouse, Rat