missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RECQL4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17008-1-AP-20UL
This item is not returnable.
View return policy
Description
This is a rabbit polyclonal antibody raised against part of C-terminal of RECQ4 of human origin. The calculated molecular weight of EIF3B is 133 kDa, but the modified EIF3B protein is about 145-150 kDa.
In humans, the RecQ helicase family includes WRN, BLM, RecQL1, RecQL4 and RecQL5 proteins, all of which contain a conserved helicase domain. WRN and BLM have been demonstrated to be the responsible genes in Werner and Bloom syndromes, respectively. RecQL1 and RecQL5 also belong to the human RecQ helicase family, but their correlation with genetic disorders, if any, is unknown. The gene encoding human RecQL4, which maps to chromosome 8q24.3, is believed to be the gene responsible for the development of Rothmund-Thomson syndrome (RTS). The levels of WRN, BLM, RecQL1, 4 and 5 are differentially up-regulated to guarantee genomic stability in cells that are transformed or actively proliferating. In humans, RecQL1 and RecQL5 map to chromosome 12p12 and 17q25, respectively.Specifications
| RECQL4 | |
| Polyclonal | |
| Unconjugated | |
| RECQL4 | |
| ATP dependent DNA helicase Q4, DNA helicase, RecQ like type 4, RecQ protein like 4, RECQ4, RECQL4, RTS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9401 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O94761 | |
| RECQL4 | |
| RECQL4 Fusion Protein Ag10707 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction