missing translation for 'onlineSavingsMsg'
Learn More

RECQL4 Antibody (2G8), Novus Biologicals™

Product Code. 18319428 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18319428 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18319428 Supplier Novus Biologicals Supplier No. H00009401M09

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RECQL4 Monoclonal antibody specifically detects RECQL4 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RECQL4
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2G8
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH13277
Gene Alias DNA helicase, RecQ-like type 4, DNA helicase, RecQ-like, type 4, EC 3.6.4.12, RecQ protein-like 4RTS, RecQ4, RECQ4ATP-dependent DNA helicase Q4
Host Species Mouse
Immunogen RECQL4 (AAH13277, 1 a.a. ~ 652 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MTRKQRESVLQKIRAAQVHVLMLTPEALVGAGGLPPAAQLPPVAFACIDEAHCLSQWSHNFRPCYLRVCKVLRERMGVHCFLGLTATATRRTASDVAQHLAVAEEPDLHGPAPVPTNLHLSVSMDRDTDQALLTLLQGKRFQNLDSIIIYCNRREDTERIAALLRTCLHAAWVPGSGGRAPKTTAEAYHAGMCSRERRRVQRAFMQGQLRVVVATVAFGMGLDRPDVRAVLHLGLPPSFESYVQAVGRAGRDGQPAHCHLFLQPQGEDLRELRRHVHADSTDFLAVKRLVQRVFPACTCTCTRPPSEQEGAVGGERPVPKYPPQEAEQLSHQAAPGPRRVCMGHERALPIQLTVQALDMPEEAIETLLCYLELHPHHWLELLATTYTHCRLNCPGGPAQLQALAHRCPPLAVCLAQQLPEDPGQGSSSVEFDMVKLVDSMGWELASVRRALCQLQWDHEPRTGVRRGTGVLVEFSELAFHLRSPGDLTAEEKDQICDFLYGRVQARERQALARLRRTFQAFHSVAFPSCGPCLEQQDEERSTRLKDLLGRYFEEEEGQEPGGMEDAQGPEPGQARLQDWEDQVRCDIRQFLSLRPEEKFSSRAVARIFHGIGSPCYPAQVYGQDRRFWRKYLHLSFHALVGLATEELLQVAR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Direct Reversal of DNA Damage
Primary or Secondary Primary
Gene ID (Entrez) 9401
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.