missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RECQ4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555263
This item is not returnable.
View return policy
Description
Immunogen sequence: WLQRCHSEVP DFLGAPKACR PDLGSEESQL LIPGESAVLG PGAGSQGPEA SAFQEVSIRV GSPQPSSSGG EKRRWNEEPW ESPAQVQQES SQAGPPSEGA GAVAVEEDPP GE Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 45%.
The protein encoded by this gene is a DNA helicase that belongs to the RecQ helicase family. DNA helicases unwind double-stranded DNA into single-stranded DNAs and may modulate chromosome segregation. This gene is predominantly expressed in thymus and testis. Mutations in this gene are associated with Rothmund-Thomson, RAPADILINO and Baller-Gerold syndromes.
Specifications
| RECQ4 | |
| Polyclonal | |
| Unconjugated | |
| RECQL4 | |
| ATP-dependent DNA helicase Q4; DNA helicase, RecQ-like type 4; DNA helicase, RecQ-like, type 4; RecQ helicase protein-like 4; RecQ helicase-like 4; RecQ like helicase 4; RecQ protein-like 4; Recq4; RECQL4; RTS | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9401 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O94761 | |
| RECQL4 | |
| Recombinant protein corresponding to Human RECQ4. Recombinant protein control fragment (Product #RP-89489). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction