missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Recoverin Recombinant Rabbit Monoclonal Antibody (4G6B4)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558187
This item is not returnable.
View return policy
Description
Immunogen sequence: MGNSKSGALS KEILEELQLN TKFSEEELCS WYQSFLKDCP TGRITQQQFQ SIYAKFFPDT DPKAYAQHVF RSFDSNLDGT LDFKEYVIAL HMTTAGKTNQ KLEWAFSLYD VDGNGTISKN EVLEIVMAIF KMITPEDVKL LPDDENTPEK RAEKIWKYFG KNDDDKLTEK EFIEGTLANK EILRLIQFEP QKVKEKMKNA.
Seems to be implicated in the pathway from retinal rod guanylate cyclase to rhodopsin. May be involved in the inhibition of the phosphorylation of rhodopsin in a calcium-dependent manner. The calcium-bound recoverin prolongs the photoresponse.
Specifications
| Recoverin | |
| Recombinant Monoclonal | |
| 0.21 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P35243 | |
| RCVRN | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human Recoverin (NP_002894.1). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 4G6B4 | |
| Unconjugated | |
| RCVRN | |
| 23 kDa photoreceptor cell-specific protein; cancer associated retinopathy antigen; cancer associated retinopathy protein; cancer-associated retinopathy protein; CAR; guanylate cyclase activator; p26; P26-calcium binding protein; Protein CAR; RCV1; RCVRN; Recoverin; S-modulin | |
| Rabbit | |
| Protein A | |
| RUO | |
| 5957 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction