missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Zika Envelope Domain-3 Protein

A cDNA sequence encoding the Zika Envelope Domain-3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13998-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15935369

  • £660.00 / 1mg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Zika Envelope Domain-3 Protein
Research Use Only
Recombinant Protein
E. coli
Lyophilized from 1 mg/ml in 20mM sodium carbonate pH 10.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Zika
His
Greater than 95.0% as determined by SDS-PAGE.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions