missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Zika Envelope Domain-3 Protein

A cDNA sequence encoding the Zika Envelope Domain-3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13998-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15925369

  • £155.00 / 100µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Zika Envelope Domain-3 Protein
Research Use Only
Recombinant Protein
E. coli
Lyophilized from 1 mg/ml in 20mM sodium carbonate pH 10.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Zika
His
Greater than 95.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers