missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Zebrafish Ephrin-B2 Fc Chimera Protein
Description
The Recombinant Zebrafish Ephrin-B2 Fc Chimera Protein is derived from NS0. The Recombinant Zebrafish Ephrin-B2 Fc Chimera Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 30219 |
| Molecular Weight (g/mol) | 48.5 kDa (monomer) |
| Name | Ephrin-B2 Fc Chimera |
| Quantity | 200 μg |
| Source | Mouse myeloma cell line,NS0-derived zebrafish Ephrin-B2 protein, (N-terminus) Zebrafish Ephrin-B2 (Leu25-Ala 222) Accession &Num; O73874 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?