missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Western Blot Protein Standards (12-90 kDa)

Product Code. 18664901 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
250 Tests
50 Tests
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18664901 250 Tests 1 set
18634970 50 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18664901 Supplier Novus Biologicals™ Supplier No. NBP311650250Tests

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Buffer 125 mM Tris-HCl, pH 6.8, 10 mM DTT, 2 mM EDTA, 2 % (W/V) SDS, 10 % (W/V) Glycerol, 0.01% (W/V) Bromophenol blue
Content And Storage Store at -20°C.
For Use With (Application) Western Blot
Quantity 250 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.