missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Western Blot Protein Standards (12-90 kDa)

Product Code. 18634970 Shop All Bio Techne Products
Click to view available options
Quantity:
250 Tests
50 Tests
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18634970

Brand: Novus Biologicals™ NBP31165050Tests

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Buffer 125 mM Tris-HCl, pH 6.8, 10 mM DTT, 2 mM EDTA, 2 % (W/V) SDS, 10 % (W/V) Glycerol, 0.01% (W/V) Bromophenol blue
Content And Storage Store at -20°C.
For Use With (Application) Western Blot
Quantity 50 Tests
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.