missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals Recombinant Virus Zika Virus VLP Suriname Z1106033 Protein
Specifications
Specifications
| For Use With (Application) | SDS-PAGE |
| Formulation | DPBS pH 7.4, 20% Sucrose. May contain traces of tartrate |
| Quantity | 0.1 mg |
| Storage Requirements | Store at -80C. Avoid freeze-thaw cycles. |
| Gene Alias | polyprotein, partial |
| Purity or Quality Grade | >95% pure by SDS-PAGE |
| Protein | Zika Virus VLP |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?