missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals Recombinant Virus Rift Valley Fever Virus Nucleocapsid ZH-548 His (N-Term) Protein
Specifications
Specifications
| For Use With (Application) | SDS-PAGE |
| Formulation | 20mM Tris pH8.0, 10mM NaCl |
| Gene ID (Entrez) | 9538291 |
| Molecular Weight (g/mol) | theroretical M.W.: 29 kDa |
| Quantity | 0.5 mg |
| Storage Requirements | Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
| Gene Alias | Fragment, NP, nucleocapsid, Nucleocapsid protein, Nucleocapsid proteinNucleoprotein, Nucleoprotein, RVFV_sS_gp2 |
| Research Category | Infections (Virus Bacteria and Parasites) |
| Purity or Quality Grade | >95% |
| Protein | Rift Valley Fever Virus Nucleocapsid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?