missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Virus Norovirus Group-1 Protein

A cDNA sequence encoding the Norovirus Group-1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12865-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15951789

  • £660.00 / 500µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Virus Norovirus Group-1 Protein
Research Use Only
Recombinant Protein
E. coli
MMMASKDATS SVDGASGAGQ LVPEVNASDP LAMDPVAGSS TAVATAGQVN PIDPWIINNF VQAPQGEFTI SPNNTPGDVL FDLSLGPHLN PFLLHLSQMY NGWVGNMRVR IMLAGNAFTA GKIIVSCIPP GFGSHNLTIA QATLFPHVIA DVRTLDPIEV PLEDVRNVLF HNNDRNQQTM RLVCMLYTPL RTGGGTGDSF VVAGRVMTCP SPDFNFLFLV PPTVEQKTRP FTLPNLPLSS LSNSRAPLPI SSMGISPDNV QSVQFQNGRC TLDGRLVGTT PVSLSHVAKI RGTSNGTVIN LTELDGTPFH PFEGPAPIGF PDLGGCDWHI NMTQFGHSSQ TQYDVDTTPD TFVPHLGSIQ ANGIGSGNYV GVLSWISPPS HPSGSQVDLW KIPNYGSSIT EATHLAPSVY PPGFGEVLVF FMSKMPGPGA YNLPCLLPQE YISHLASEQA PTVGEAALLH YVDPDTGRNL GEFKAYPDGF LTCVPNGASS GPQQLPINGV FVFVSWVSRF YQLKPVGTAS SARGRLGLRR
Protein is >95% pure as determined by 12% PAGE (coomassie staining).
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Virus
His
Phosphate buffer with 0.05% NaN3.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers