missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Virus Norovirus Group-1 P-Domain Protein
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
Specifications
Specifications
| Name | Virus Norovirus Group-1 P-Domain Protein |
| Quantity | 100 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Virus |
| Species | E. coli |
| Protein Tag | His |
| Buffer | Phosphate buffer and 10 mM K2CO3. |
| Purity or Quality Grade | Protein is >95% pure as determined by 12% PAGE (coomassie staining). |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction