missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals Recombinant Virus Dengue Virus 4 Envelope Dominica/814669/1981 His (C-Term) Protein
Shop All Bio Techne ProductsSpecifications
Specifications
| For Use With (Application) | SDS-PAGE |
| Formulation | 20mM Tris-HCl, 110mM sodium chloride, pH7.8. |
| Quantity | 0.1 mg |
| Storage Requirements | Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
| Purity or Quality Grade | >95% pure by SDS-PAGE |
| Protein | Dengue Virus 4 Envelope |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?