missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals Recombinant Virus Dengue Virus 3 Envelope Sri Lanka D3/H/IMTSSA-SRI/2000/1266 His (C-Term) Protein
Specifications
Specifications
| For Use With (Application) | SDS-PAGE |
| Formulation | 20mM Tris-HCl, 185mM sodium chloride, pH7.8. |
| Molecular Weight (g/mol) | theroretical M.W.: 45 kDa |
| Quantity | 0.1 mg |
| Storage Requirements | Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
| Gene Alias | polyprotein |
| Purity or Quality Grade | >95% |
| Protein | Dengue Virus 3 Envelope |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?