missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals Recombinant Virus Dengue Virus 2 Envelope His (C-Term) Protein

Product Code. p-7229490 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.10mg
0.5mg
This item is not returnable. View return policy

Product Code. 18340636

Brand: Novus Biologicals NBP3148120.1MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl, 110mM sodium chloride, pH7.8.
Quantity 0.1 mg
Storage Requirements Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Gene Alias Envelope protein E
Protein Dengue Virus 2 Envelope
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.