missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals Recombinant Virus Chikungunya Virus E2 Fc Protein

Product Code. 18334582 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.1mg
0.50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18334582 0.1 mg 0.1mg
18325714 0.5 mg 0.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18334582 Supplier Novus Biologicals Supplier No. NBP3148180.1MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Concentration 0.32 mg/ml
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM HEPES, pH7.0, 300mM NaCl, 0.25% Triton X-100
Molecular Weight (g/mol) theroretical M.W.: 69 kDa
Quantity 0.1 mg
Storage Requirements Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Gene Alias Chikungunya, E2 envelope glycoprotein, P130, Spike glycoprotein E2
Purity or Quality Grade >95%
Protein Chikungunya Virus E2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.