missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Viral Monkeypox Virus A29L (HEK-293-expressed) His (C-Term) Protein

Product Code. 18354643 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354643 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18354643 Supplier Novus Biologicals™ Supplier No. NBP318267

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration LYOPH
For Use With (Application) SDS-PAGE
Formulation Lyophilized from a 0.22 um filtered solution of PBS, pH 7.4
Quantity 100 μg
Storage Requirements Store at -20°C.
Endotoxin Concentration <0.1 EU/μg
Gene Symbol A29L
Reconstitution Reconstitute to a concentration of 0.1-0.5 mg/mL in sterile distilled water.
Purity or Quality Grade >95%, by SDS-PAGE
Protein Monkeypox Virus A29L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.