missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Viral CMV UL146/vCXC1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 620-CM-025
Additional Details : Weight : 0.00970kg
Description
The Recombinant Viral CMV UL146/vCXC1 Protein is derived from E. coli. The Recombinant Viral CMV UL146/vCXC1 Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. | |
CMV UL146/vCXC1 | |
E. coli-derived viral UL146/vCXC1 protein Thr23-Gly117 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
11.1 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |