missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Vespula vulgaris 1.0101 Strep (N-Term) Protein
Shop All Bio Techne ProductsSpecifications
Specifications
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | Lyophilized from a 0.22 um filtered solution in Tris buffer (100 mM Tris, 150 mM NaCl, pH 8), preservative- and carrier-free. |
| Quantity | 0.1 mg |
| Storage Requirements | Store at -20 to -80C. Avoid freeze-thaw cycles. |
| Gene Alias | Allergen Ves v I, EC:3.1.1.32, EC:3.1.1.4, Phospholipase A1, Ves v 1 |
| Reconstitution | Reconstitute with sterile deionized water with or without azide. |
| Form | Lyophilized |
| Purity or Quality Grade | >95%, by HPLC. |
| Protein | Vespula vulgaris 1.0101 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?