missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Toxoplasma Gondii T. gondii p40 Protein

A cDNA sequence encoding the T. gondii p40 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13667-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15964319

  • £170.83 / 100µg
Estimated Shipment: 24-02-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Toxoplasma Gondii T. gondii p40 Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Toxoplasma Gondii
His
Toxoplasma protein is >95% pure as determined by 10% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
1x PBS and 25mM arginine.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers